siRNA Details
Gene Symbol: C1orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030806.3 | NP_110433.1 | 4 | 688 | ||
GenBank mRNA | AF312864.1 | - | 4 | 610 | ||
AK056487.1 | - | 4 | 424 | |||
AK312062.1 | - | 3 | 325 | |||
AY258286.1 | - | 4 | 601 | |||
BC028567.2 | - | 4 | 646 | 3635521 |
