siRNA Details
Gene Symbol: TSPAN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030927.2 | NP_112189.2 | 5 | 520 | ||
XM_006717992.3 | XP_006718055.1 | 5 | 676 | |||
XM_011540219.2 | XP_011538521.1 | 7 | 710 | |||
XM_011540220.2 | XP_011538522.1 | 7 | 632 | |||
XM_011540221.2 | XP_011538523.1 | 6 | 494 | |||
XM_011540222.2 | XP_011538524.1 | 6 | 672 | |||
XM_017016707.1 | XP_016872196.1 | 7 | 668 | |||
XM_017016708.1 | XP_016872197.1 | 6 | 629 | |||
XM_017016709.1 | XP_016872198.1 | 6 | 677 | |||
XM_017016710.1 | XP_016872199.1 | 6 | 628 | |||
XM_017016711.1 | XP_016872200.1 | 6 | 493 | |||
XM_017016712.1 | XP_016872201.1 | 6 | 696 | |||
XM_017016713.1 | XP_016872202.1 | 5 | 516 | |||
GenBank mRNA | AF311903.1 | - | 5 | 485 | ||
AL136638.1 | - | 5 | 491 | |||
BC002920.2 | - | 4 | 415 | 3954042 | ||
BC093016.1 | - | 5 | 520 | 30528940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
