siRNA Details
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 6, 7 | 988 | ||
NM_001160225.1 | NP_001153697.1 | 6, 7 | 637 | |||
NM_030954.3 | NP_112216.3 | 6, 7 | 758 | |||
NR_027668.1 | 5, 6 | 914 | ||||
NR_027669.1 | 5, 6 | 684 | ||||
XM_006716404.2 | XP_006716467.1 | 7, 8 | 745 | |||
XM_006716405.2 | XP_006716468.1 | 4, 5 | 504 | |||
XM_011544666.2 | XP_011542968.1 | 6, 7 | 542 | |||
XM_017013881.1 | XP_016869370.1 | 7, 8 | 685 | |||
XM_017013882.1 | XP_016869371.1 | 5, 6 | 530 | |||
GenBank mRNA | AF209504.1 | - | 1, 2 | 95 | ||
AK027748.1 | - | 6, 7 | 674 | |||
AK090864.1 | - | 5, 6 | 847 | |||
AK225086.1 | - | 6, 7 | 638 | |||
AK225568.1 | - | 6, 7 | 654 | |||
BC013422.1 | - | 5, 6 | 549 | |||
BC039461.1 | - | 6, 7 | 745 | |||
BC058289.1 | - | 6, 7 | 621 |
