siRNA Details
Gene Symbol: KRTAP1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030967.2 | NP_112229.1 | 1 | 258 | ||
GenBank mRNA | AB052868.1 | - | 2 | 120 | ||
AB052934.1 | - | 1 | 258 | |||
AJ406926.1 | - | 1 | 227 | |||
BC069321.1 | - | 1 | 244 | |||
BC069450.1 | - | 1 | 244 | |||
BC105069.1 | - | 1 | 233 | 8144072 | ||
BC105071.1 | - | 1 | 233 | 8144074 |
