siRNA Details
Gene Symbol: DRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130090.1 | NP_001123562.1 | 14 | 1776 | ||
NM_031294.3 | NP_112584.3 | 13 | 1687 | |||
XM_006721583.3 | XP_006721646.1 | 11 | 1471 | |||
XM_011524018.2 | XP_011522320.1 | 14 | 1836 | |||
XM_011524019.2 | XP_011522321.1 | 14 | 1682 | |||
XM_011524020.2 | XP_011522322.1 | 14 | 1961 | |||
XM_011524021.2 | XP_011522323.1 | 14 | 1777 | |||
XM_011524025.2 | XP_011522327.1 | 8 | 2103 | |||
XM_011524026.2 | XP_011522328.1 | 8 | 878 | |||
GenBank mRNA | AK093317.1 | - | 12 | 1538 | ||
AL136926.1 | - | 10 | 5443 | |||
BC047873.1 | - | 13 | 1615 | 5760028 | ||
BC050419.1 | - | 14 | 1637 | 5766999 | ||
BC050665.1 | - | 14 | 1633 | 5203312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
