siRNA Details
Gene Symbol: DRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130090.1 | NP_001123562.1 | 14, 15 | 1829 | ||
NM_031294.3 | NP_112584.3 | 13, 14 | 1740 | |||
XM_006721583.3 | XP_006721646.1 | 11, 12 | 1524 | |||
XM_011524018.2 | XP_011522320.1 | 14, 15 | 1889 | |||
XM_011524019.2 | XP_011522321.1 | 14, 15 | 1735 | |||
XM_011524020.2 | XP_011522322.1 | 14, 15 | 2014 | |||
XM_011524021.2 | XP_011522323.1 | 14, 15 | 1830 | |||
XM_011524025.2 | XP_011522327.1 | 8, 9 | 2156 | |||
XM_011524026.2 | XP_011522328.1 | 8, 9 | 931 | |||
GenBank mRNA | AK093317.1 | - | 12, 13 | 1591 | ||
AL136926.1 | - | 10, 11 | 5496 | |||
BC047873.1 | - | 13, 14 | 1668 | 5760028 | ||
BC050419.1 | - | 14, 15 | 1690 | 5766999 | ||
BC050665.1 | - | 14, 15 | 1686 | 5203312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
