siRNA Details
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031725.5 | NP_001026895.2 | 6 | 1677 | ||
NM_001320181.1 | NP_001307110.1 | 6 | 1677 | |||
NM_001320182.1 | NP_001307111.1 | 3 | 1103 | |||
GenBank mRNA | AK299219.1 | - | 6 | 1548 | ||
AK307569.1 | - | 1605 | ||||
AL136611.1 | - | 6 | 1590 | |||
BC041801.1 | - | 6 | 1668 | 5302098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
