siRNA Details
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 5 | 564 | ||
XM_005254708.4 | XP_005254765.1 | 8 | 932 | |||
XM_005254710.4 | XP_005254767.1 | 9 | 1070 | |||
XM_005254711.4 | XP_005254768.1 | 8 | 1292 | |||
XM_005254712.4 | XP_005254769.1 | 8 | 949 | |||
XM_005254713.4 | XP_005254770.1 | 8 | 1846 | |||
XM_005254714.3 | XP_005254771.1 | 7 | 835 | |||
XM_005254715.2 | XP_005254772.1 | 7 | 1541 | |||
XM_006720717.3 | XP_006720780.1 | 8 | 1666 | |||
XM_017022665.1 | XP_016878154.1 | 8 | 2017 | |||
XM_017022666.1 | XP_016878155.1 | 8 | 876 | |||
XM_017022667.1 | XP_016878156.1 | 6 | 738 | |||
XM_017022668.1 | XP_016878157.1 | 8 | 1665 | |||
XM_017022669.1 | XP_016878158.1 | 8 | 1666 | |||
GenBank mRNA | AF402000.1 | - | 5 | 564 |
