siRNA Details
Gene Symbol: CYP2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000764.2 | NP_000755.2 | 7 | 1662 | ||
NM_030589.2 | NP_085079.2 | 6 | 1509 | |||
GenBank mRNA | BC117165.1 | - | 7 | 1165 | 40125716 | |
BC143325.1 | - | 7 | 1165 | 9051830 |
