siRNA Details
Gene Symbol: H2AFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080720.1 | NP_542451.1 | 1 | 481 | ||
GenBank mRNA | AF254576.1 | - | 1 | 481 |
Gene Symbol: H2AFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017990.1 | NP_001017990.1 | 1 | 481 |
Gene Symbol: H2AFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017991.2 | NP_001017991.1 | 1 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
