siRNA Details
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB042647.1 | - | 5, 6 | 579 | ||
AK054702.1 | - | 5, 6 | 649 | |||
BC029860.1 | - | 5, 6 | 602 | 5171860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
