siRNA Details
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000772.2 | NP_000763.1 | 5 | 1105 | ||
GenBank mRNA | AK223510.1 | - | 5 | 804 | ||
AK308558.1 | - | 5 | 760 | |||
AK313403.1 | - | 5 | 804 | |||
BC069666.1 | - | 5 | 973 | 7262569 | ||
BC096257.1 | - | 5 | 824 | 40006072 | ||
BC096258.1 | - | 5 | 824 | 40006073 | ||
M61853.1 | - | 5 | 821 | |||
M61856.1 | - | 5 | 978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
