siRNA Details
Gene Symbol: HORMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199829.1 | NP_001186758.1 | 8 | 512 | ||
NM_032132.4 | NP_115508.2 | 9 | 533 | |||
XM_006711570.2 | XP_006711633.1 | 8 | 427 | |||
XM_006711571.2 | XP_006711634.1 | 5 | 289 | |||
XM_011510053.2 | XP_011508355.1 | 9 | 601 | |||
XM_011510054.2 | XP_011508356.1 | 9 | 555 | |||
XM_011510055.2 | XP_011508357.1 | 8 | 351 | |||
XM_011510056.2 | XP_011508358.1 | 4 | 133 | |||
GenBank mRNA | AK097390.1 | - | 9 | 521 | ||
AK098633.1 | - | 9 | 491 | |||
AL136755.2 | - | 8 | 509 | |||
AY626344.1 | - | 8 | 509 | |||
BC033014.1 | - | 7 | 434 | 4827705 | ||
BC047406.1 | - | 9 | 521 | 5298160 | ||
CR533505.1 | - | 7 | 394 |
