siRNA Details
Gene Symbol: ELMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135021.1 | NP_001128493.1 | 10 | 1127 | ||
NM_001135022.1 | NP_001128494.1 | 9 | 943 | |||
NM_001135023.1 | NP_001128495.1 | 8 | 899 | |||
NM_032213.4 | NP_115589.2 | 7 | 804 | |||
XM_005264596.3 | XP_005264653.1 | 9 | 943 | |||
XM_005264597.3 | XP_005264654.1 | 8 | 1225 | |||
XM_005264598.3 | XP_005264655.1 | 8 | 848 | |||
XM_017005078.1 | XP_016860567.1 | 8 | 799 | |||
XM_017005079.1 | XP_016860568.1 | 7 | 755 | |||
GenBank mRNA | AF258573.1 | - | 6 | 581 | ||
AK307252.1 | - | 5 | 686 | |||
AL713718.1 | - | 5 | 361 | |||
BC001942.1 | - | 7 | 721 | 2819402 | ||
BC010991.2 | - | 9 | 875 | 3936410 | ||
BC018666.2 | - | 7 | 721 | 2819402 | ||
BC054504.1 | - | 7 | 2671 | |||
BC112324.1 | - | 8 | 862 | 5200867 | ||
DQ256726.1 | - | 6 | 470 | |||
DQ256727.1 | - | 6 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
