siRNA Details
Gene Symbol: TMEM175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297423.1 | NP_001284352.1 | 6, 7 | 1023 | ||
NM_001297424.1 | NP_001284353.1 | 4, 5 | 339 | |||
NM_001297425.1 | NP_001284354.1 | 4, 5 | 336 | |||
NM_001297426.1 | NP_001284355.1 | 7, 8 | 803 | |||
NM_001297427.1 | NP_001284356.1 | 6, 7 | 1059 | |||
NM_001297428.1 | NP_001284357.1 | 4, 5 | 375 | |||
NM_032326.3 | NP_115702.1 | 6, 7 | 562 | |||
XM_005272303.4 | XP_005272360.1 | 7, 8 | 719 | |||
XM_005272304.1 | XP_005272361.1 | 6, 7 | 528 | |||
XM_005272307.4 | XP_005272364.1 | 6, 7 | 488 | |||
XM_017008701.1 | XP_016864190.1 | 7, 8 | 639 | |||
XM_017008702.1 | XP_016864191.1 | 4, 5 | 286 | |||
XM_017008703.1 | XP_016864192.1 | 7, 8 | 1081 | |||
XM_017008705.1 | XP_016864194.1 | 3, 4 | 231 | |||
GenBank mRNA | AK090848.1 | - | 6, 7 | 953 | ||
AK309132.1 | - | 6, 7 | 488 | |||
AL834199.1 | - | 7, 8 | 673 | |||
BC005158.2 | - | 6, 7 | 475 | 3505819 | ||
BI819231.1 | - | 4, 5 | 283 | |||
CB992640.1 | - | 4, 5 | 304 | |||
CF242841.1 | - | 4, 5 | 278 |
