siRNA Details
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032331.3 | NP_115707.2 | 3 | 548 | ||
GenBank mRNA | BC005835.2 | - | 3 | 544 | ||
BC012449.1 | - | 3 | 521 | |||
BC069005.1 | - | 3 | 534 | |||
BU568980.1 | - | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
