siRNA Details
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128612.2 | NP_001122084.1 | 5 | 805 | ||
NM_001128613.2 | NP_001122085.1 | 4 | 820 | |||
NM_001271831.1 | NP_001258760.1 | 4 | 706 | |||
NM_032344.3 | NP_115720.1 | 5 | 919 | |||
GenBank mRNA | BC006129.1 | - | 5 | 903 | 3625545 | |
BM923060.1 | - | 4 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
