siRNA Details
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128612.2 | NP_001122084.1 | 3 | 597 | ||
NM_032344.3 | NP_115720.1 | 3 | 711 | |||
GenBank mRNA | BC006129.1 | - | 3 | 695 | 3625545 | |
BG705568.1 | - | 3 | 602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
