siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032433.2 | NP_115809.1 | 12 | 1112 | ||
XM_005260098.3 | XP_005260155.1 | 13 | 1425 | |||
XM_006722924.2 | XP_006722987.1 | 12 | 1189 | |||
XM_011528362.2 | XP_011526664.1 | 12 | 1153 | |||
XM_011528363.2 | XP_011526665.1 | 14 | 2253 | |||
XM_011528365.2 | XP_011526667.1 | 14 | 1594 | |||
XM_011528366.2 | XP_011526668.1 | 13 | 1422 | |||
XM_011528367.2 | XP_011526669.1 | 13 | 1435 | |||
XM_011528368.2 | XP_011526670.1 | 13 | 1342 | |||
XM_011528369.2 | XP_011526671.1 | 13 | 1355 | |||
XM_011528371.2 | XP_011526673.1 | 6 | 1519 | |||
XM_011528372.2 | XP_011526674.1 | 5 | 448 | |||
XM_017027367.1 | XP_016882856.1 | 12 | 1129 | |||
XM_017027368.1 | XP_016882857.1 | 11 | 997 | |||
XM_017027369.1 | XP_016882858.1 | 13 | 2750 | |||
XM_017027370.1 | XP_016882859.1 | 13 | 2763 | |||
XM_017027371.1 | XP_016882860.1 | 13 | 1375 | |||
GenBank mRNA | AB058709.1 | - | 5 | 426 | ||
AF372702.1 | - | 12 | 1069 | |||
AK094028.1 | - | 13 | 1355 | |||
AK122817.1 | - | 5 | 993 | |||
AL832544.1 | - | 6 | 604 | |||
BX647475.1 | - | 12 | 1073 |
