siRNA Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032483.3 | NP_115872.2 | 7 | 763 | ||
GenBank mRNA | AF212238.1 | - | 7 | 717 | ||
BC106014.1 | - | 7 | 733 | 3877263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
