siRNA Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102559.1 | NP_001096029.1 | 5, 6 | 476 | ||
NM_032483.3 | NP_115872.2 | 5, 6 | 476 | |||
XM_005273661.4 | XP_005273718.2 | 5, 6 | 670 | |||
XM_011544671.1 | XP_011542973.1 | 5, 6 | 460 | |||
XM_011544672.1 | XP_011542974.1 | 5, 6 | 816 | |||
XM_017013907.1 | XP_016869396.1 | 5, 6 | 816 | |||
GenBank mRNA | AF212238.1 | - | 5, 6 | 430 | ||
AK292118.1 | - | 5, 6 | 401 | |||
BC033025.1 | - | 4, 5 | 536 | 5267610 | ||
BC106014.1 | - | 5, 6 | 446 | 3877263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
