siRNA Details
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045128.1 | 10 | 1369 | |||
GenBank mRNA | AF332225.1 | - | 4 | 645 | ||
AL832583.2 | - | 5 | 4915 | |||
CD242693.1 | - | 5 | 712 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045128.1 | 10 | 1369 | |||
GenBank mRNA | AF332225.1 | - | 4 | 645 | ||
AL832583.2 | - | 5 | 4915 | |||
CD242693.1 | - | 5 | 712 |