siRNA Details
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 6, 7 | 792 | ||
XM_011531482.2 | XP_011529784.1 | 6, 7 | 830 | |||
XM_011531483.1 | XP_011529785.1 | 5, 6 | 452 | |||
GenBank mRNA | AF271087.1 | - | 1, 2 | 139 | ||
AF271088.1 | - | 3, 4 | 267 | |||
BC114927.1 | - | 6, 7 | 623 | 40028543 |
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 6, 7 | 807 | ||
XM_005262602.1 | XP_005262659.1 | 6, 7 | 746 | |||
XM_011531509.2 | XP_011529811.1 | 6, 7 | 833 | |||
GenBank mRNA | AF248482.1 | - | 6, 7 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
