siRNA Details
Gene Symbol: ZBTB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316978.1 | NP_001303907.1 | 3 | 1865 | ||
NM_001316979.1 | NP_001303908.1 | 3 | 1592 | |||
NM_001316980.1 | NP_001303909.1 | 3 | 1415 | |||
NM_001316981.1 | NP_001303910.1 | 3 | 1717 | |||
NM_001316982.1 | NP_001303911.1 | 3 | 1535 | |||
NM_032792.3 | NP_116181.1 | 3 | 1621 | |||
XM_006723445.3 | XP_006723508.1 | 3 | 1635 | |||
GenBank mRNA | AK027392.1 | - | 3 | 1404 | ||
AL831909.1 | - | 2 | 166 | |||
BC024738.1 | - | 3 | 1616 | 5110354 |
