siRNA Details
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204817.1 | NP_001191746.1 | 3 | 2659 | ||
NM_032828.3 | NP_116217.1 | 3 | 2662 | |||
GenBank mRNA | AK027616.1 | - | 4 | 2303 | ||
AK160374.1 | - | 2 | 2538 | |||
BC126237.1 | - | 3 | 2524 | 8991953 | ||
BC126239.1 | - | 3 | 2525 | 8991955 |
