siRNA Details
Gene Symbol: KRTAP9-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318227.1 | NP_001305156.1 | 2 | 856 | ||
NM_030975.2 | NP_112237.2 | 1 | 841 | |||
GenBank mRNA | AJ406949.1 | - | 1 | 667 | ||
AJ406951.1 | - | 3 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
