siRNA Details
Gene Symbol: KIAA1671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145206.1 | NP_001138678.1 | 3 | 2205 | ||
XM_005261793.2 | XP_005261850.1 | 5 | 2518 | |||
XM_005261794.4 | XP_005261851.1 | 5 | 2471 | |||
XM_006724346.2 | XP_006724409.1 | 6 | 2457 | |||
GenBank mRNA | BC171801.1 | - | 3 | 2205 | 9054455 |
