siRNA Details
Gene Symbol: ITPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272012.1 | NP_001258941.1 | 2 | 1327 | ||
NM_001272013.1 | NP_001258942.1 | 2 | 1447 | |||
NM_033397.3 | NP_203755.1 | 3 | 1639 | |||
XM_005270257.2 | XP_005270314.1 | 1 | 1203 | |||
GenBank mRNA | AB051541.1 | - | 2 | 1358 | ||
AK055608.1 | - | 1 | 50 | |||
AK056526.1 | - | 2 | 1259 | |||
AK094802.1 | - | 1 | 679 | |||
AK129877.1 | - | 3 | 1350 | |||
BC040536.1 | - | 2 | 1299 | 5265379 | ||
BC070108.1 | - | 3 | 1639 | 4837204 |
