siRNA Details
Gene Symbol: DPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080650.3 | NP_542381.1 | 4 | 422 | ||
XM_011522160.1 | XP_011520462.1 | 4 | 441 | |||
XM_017022707.1 | XP_016878196.1 | 4 | 441 | |||
XM_017022708.1 | XP_016878197.1 | 4 | 441 | |||
XM_017022709.1 | XP_016878198.1 | 4 | 441 | |||
GenBank mRNA | AK094007.1 | - | 4 | 382 | ||
BC008485.2 | - | 3 | 338 | |||
BC066652.1 | - | 4 | 369 | 6066017 | ||
DB250468.1 | - | 4 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
