siRNA Details
Gene Symbol: ZNF835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005850.2 | NP_001005850.2 | 2 | 1446 | ||
XM_005259382.2 | XP_005259439.1 | 2 | 1449 | |||
XM_005259383.3 | XP_005259440.1 | 2 | 1481 | |||
GenBank mRNA | AK023017.1 | - | 1 | 1058 | ||
AK299565.1 | - | 2 | 1446 |
