siRNA Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281746.1 | NP_001268675.1 | 2 | 276 | ||
NM_001281747.1 | NP_001268676.1 | 2 | 258 | |||
NM_138569.2 | NP_612636.2 | 2 | 276 | |||
XM_005249476.4 | XP_005249533.1 | 2 | 229 | |||
XM_005249480.4 | XP_005249537.1 | 2 | 229 | |||
XM_006715245.3 | XP_006715308.2 | 2 | 368 | |||
XM_017011465.1 | XP_016866954.1 | 2 | 365 | |||
XM_017011466.1 | XP_016866955.1 | 2 | 362 | |||
XM_017011467.1 | XP_016866956.1 | 2 | 371 | |||
XM_017011468.1 | XP_016866957.1 | 2 | 370 | |||
XM_017011469.1 | XP_016866958.1 | 2 | 162 | |||
XM_017011470.1 | XP_016866959.1 | 2 | 460 | |||
GenBank mRNA | AK055530.1 | - | 2 | 201 | ||
AK294890.1 | - | 2 | 229 | |||
AK311190.1 | - | 2 | 229 | |||
AK316157.1 | - | 2 | 209 | |||
BX091237.1 | - | 2 | 143 | |||
BX647567.1 | - | 2 | 222 | |||
DA761851.1 | - | 2 | 276 | |||
KU178823.1 | - | 2 | 133 | |||
KU178824.1 | - | 2 | 133 | |||
KU178825.1 | - | 2 | 133 | |||
KU178826.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
