siRNA Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055530.1 | - | 8, 9 | 986 | ||
AK294890.1 | - | 9, 10 | 2586 | |||
AK311190.1 | - | 9, 10 | 2586 | |||
BC009010.1 | - | 3, 4 | 526 | 4158300 | ||
BX113168.1 | - | 1, 2 | 64 | |||
BX647567.1 | - | 9, 10 | 2579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
