siRNA Details
Gene Symbol: ZNF486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052852.3 | NP_443084.2 | 4 | 1029 | ||
GenBank mRNA | AB209550.1 | - | 2 | 732 | ||
BC027608.1 | - | 4 | 1034 | |||
BC117268.1 | - | 4 | 969 | 40125819 |
