siRNA Details
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304458.1 | NP_001291387.1 | 10, 11 | 2424 | ||
NM_138364.3 | NP_612373.2 | 11, 12 | 2573 | |||
XM_017008823.1 | XP_016864312.1 | 11, 12 | 2314 | |||
XM_017008824.1 | XP_016864313.1 | 10, 11 | 2246 | |||
XM_017008825.1 | XP_016864314.1 | 10, 11 | 2300 | |||
XM_017008826.1 | XP_016864315.1 | 9, 10 | 2195 | |||
GenBank mRNA | AK096703.1 | - | 10, 11 | 2319 | ||
AK128483.1 | - | 5, 6 | 1656 | |||
AK292904.1 | - | 11, 12 | 2506 | |||
AL137452.1 | - | 2, 3 | 299 | |||
BC004337.1 | - | 3, 4 | 852 | |||
BC021250.2 | - | 11, 12 | 2328 | |||
BC064403.1 | - | 11, 12 | 2472 | 5547611 |
