siRNA Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178167.3 | NP_835461.2 | 9 | 1617 | ||
GenBank mRNA | AK024487.1 | - | 6 | 2476 | ||
AL834428.1 | - | 3 | 401 | |||
BC010990.2 | - | 5 | 812 | |||
BC041015.1 | - | 3 | 402 | |||
BC050477.1 | - | 9 | 1558 | 6055142 |
