siRNA Details
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002862.2 | NP_001002862.1 | 7 | 1395 | ||
NM_198440.3 | NP_940842.2 | 6 | 1496 | |||
GenBank mRNA | AK092582.1 | - | 1 | 540 | ||
AK096976.1 | - | 5 | 1407 | |||
AK125830.1 | - | 5 | 1697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
