siRNA Details
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 5 | 932 | ||
NM_152267.3 | NP_689480.2 | 7 | 1100 | |||
NR_024209.1 | 7 | 1085 | ||||
NR_024210.1 | 7 | 1267 | ||||
NR_024211.1 | 6 | 987 | ||||
NR_024212.1 | 6 | 876 | ||||
GenBank mRNA | AK027035.1 | - | 6 | 798 | ||
AK075154.1 | - | 1 | 182 | |||
AK095947.1 | - | 5 | 841 | |||
AK291236.1 | - | 7 | 1009 | |||
AK297414.1 | - | 6 | 957 | |||
BC026040.1 | - | 7 | 992 | 4866294 | ||
BC033166.1 | - | 7 | 1035 | 4549320 | ||
BC035684.1 | - | 7 | 1059 | 5750180 | ||
BX648019.1 | - | 7 | 1009 | |||
DQ296559.1 | - | 7 | 1041 | |||
DQ296561.1 | - | 7 | 1041 | |||
DQ296562.1 | - | 5 | 873 | |||
DQ296564.1 | - | 7 | 1209 | |||
DQ296565.1 | - | 7 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
