siRNA Details
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 5 | 1162 | ||
NM_152267.3 | NP_689480.2 | 7 | 1330 | |||
NR_024209.1 | 7 | 1315 | ||||
NR_024210.1 | 7 | 1497 | ||||
NR_024211.1 | 6 | 1217 | ||||
NR_024212.1 | 6 | 1106 | ||||
GenBank mRNA | AK027035.1 | - | 6 | 1028 | ||
AK075154.1 | - | 1 | 412 | |||
AK095947.1 | - | 5 | 1071 | |||
BC017302.1 | - | 1 | 226 | |||
BC026040.1 | - | 7 | 1222 | 4866294 | ||
BC033166.1 | - | 7 | 1265 | 4549320 | ||
BC035684.1 | - | 7 | 1289 | 5750180 | ||
BX648019.1 | - | 7 | 1239 | |||
DQ296559.1 | - | 7 | 1271 | |||
DQ296562.1 | - | 5 | 1103 | |||
DQ296564.1 | - | 7 | 1439 | |||
DQ296565.1 | - | 7 | 1271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
