siRNA Details
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 2 | 171 | ||
NM_004403.2 | NP_004394.1 | 2 | 416 | |||
GenBank mRNA | AF007790.2 | - | 1 | 71 | ||
AF073308.1 | - | 2 | 140 | |||
AK094714.1 | - | 4 | 742 | |||
AK290775.1 | - | 2 | 147 | |||
AK314402.1 | - | 2 | 156 | |||
BC099911.1 | - | 2 | 132 | 5287362 | ||
BC125065.1 | - | 2 | 138 | 40117291 | ||
DA197867.1 | - | 2 | 383 | |||
DA820184.1 | - | 2 | 171 | |||
DA908548.1 | - | 2 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
