siRNA Details
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105543.1 | NP_001099013.1 | 2, 3 | 1418 | ||
NM_020911.1 | NP_065962.1 | 2, 3 | 1418 | |||
NM_181775.3 | NP_861440.2 | 3, 4 | 1566 | |||
XM_005250686.4 | XP_005250743.1 | 2, 3 | 1816 | |||
XM_006716171.3 | XP_006716234.1 | 3, 4 | 1909 | |||
XM_011516676.2 | XP_011514978.1 | 2, 3 | 1324 | |||
XM_017012779.1 | XP_016868268.1 | 2, 3 | 1324 | |||
GenBank mRNA | AK123428.1 | - | 2, 3 | 1324 | ||
AL137352.1 | - | 1, 2 | 575 | |||
AY358850.1 | - | 1, 2 | 611 | |||
BC028744.2 | - | 3, 4 | 1566 |
