siRNA Details
Gene Symbol: TDRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527471.2 | XP_011525773.1 | 23 | 2776 | ||
XM_011527472.2 | XP_011525774.1 | 23 | 2761 | |||
XM_011527473.2 | XP_011525775.1 | 23 | 2776 | |||
XM_011527474.2 | XP_011525776.1 | 22 | 2656 | |||
XM_011527475.2 | XP_011525777.1 | 23 | 2776 | |||
XM_011527476.2 | XP_011525778.1 | 23 | 2776 | |||
XM_017027458.1 | XP_016882947.1 | 23 | 2776 | |||
XM_017027459.1 | XP_016882948.1 | 13 | 1759 | |||
GenBank mRNA | BC049000.1 | - | 23 | 3001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
