siRNA Details
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 13 | 1603 | ||
XM_011534216.2 | XP_011532518.1 | 13 | 1701 | |||
XM_017007461.1 | XP_016862950.1 | 14 | 1787 | |||
GenBank mRNA | AJ318099.1 | - | 13 | 1587 | ||
BC006846.1 | - | 4 | 517 | |||
BC043595.1 | - | 13 | 1596 | 5769824 |
