siRNA Details
Gene Symbol: ZNF585B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152279.3 | NP_689492.3 | 5 | 888 | ||
GenBank mRNA | AK091586.1 | - | 2 | 62 | ||
AK094989.1 | - | 1 | 890 | |||
AY147015.1 | - | 5 | 890 | |||
BC080550.1 | - | 6 | 653 | |||
BC093625.1 | - | 4 | 730 | 7939470 |
