siRNA Details
Gene Symbol: TMEM132C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136103.2 | NP_001129575.2 | 2 | 366 | ||
XM_011538998.2 | XP_011537300.1 | 2 | 692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
