siRNA Details
Gene Symbol: NAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128931.1 | NP_001122403.1 | 4, 5 | 914 | ||
NM_138386.2 | NP_612395.2 | 4, 5 | 914 | |||
XM_011532410.2 | XP_011530712.1 | 4, 5 | 4381 | |||
XM_011532411.2 | XP_011530713.1 | 5, 6 | 988 | |||
GenBank mRNA | AK097352.1 | - | 4, 5 | 824 | ||
AK308601.1 | - | 4, 5 | 883 | |||
BC008207.1 | - | 4, 5 | 914 | 3447276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
