siRNA Details
Gene Symbol: NAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138386.2 | NP_612395.2 | 8 | 1292 | ||
XM_011532410.2 | XP_011530712.1 | 8 | 4759 | |||
XM_011532411.2 | XP_011530713.1 | 9 | 1366 | |||
GenBank mRNA | BC008207.1 | - | 8 | 1292 | 3447276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
