siRNA Details
Gene Symbol: SLC38A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033518.3 | NP_277053.2 | 15 | 1324 | ||
XM_005272694.3 | XP_005272751.2 | 14 | 1314 | |||
XM_005272695.4 | XP_005272752.3 | 14 | 1316 | |||
XM_005272697.2 | XP_005272754.2 | 14 | 1267 | |||
XM_005272698.4 | XP_005272755.2 | 14 | 1205 | |||
XM_006724569.3 | XP_006724632.1 | 15 | 1324 | |||
XM_017029960.1 | XP_016885449.1 | 14 | 1236 | |||
XM_017029961.1 | XP_016885450.1 | 14 | 1208 | |||
GenBank mRNA | AF276889.1 | - | 14 | 1989 | ||
AF318358.1 | - | 11 | 1435 | |||
AK094788.1 | - | 14 | 1243 | |||
BC019246.2 | - | 14 | 1174 | 3506044 | ||
BC027721.2 | - | 10 | 865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
