siRNA Details
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144576.3 | NP_653177.3 | 1, 2 | 393 | ||
GenBank mRNA | AK057003.1 | - | 1, 2 | 309 | ||
AY358861.1 | - | 1, 2 | 81 | |||
BC047444.1 | - | 1, 2 | 121 | |||
BC073923.1 | - | 1, 2 | 277 | 5188138 | ||
DB271789.1 | - | 1, 2 | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
