siRNA Details
Gene Symbol: DMKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035516.3 | NP_001030593.1 | 2, 3 | 135 | ||
NM_001126056.2 | NP_001119528.2 | 14, 15 | 1435 | |||
NM_001126059.2 | NP_001119531.1 | 8, 9 | 486 | |||
NM_001308383.1 | NP_001295312.1 | 7, 8 | 456 | |||
NM_033317.4 | NP_201574.3 | 12, 13 | 1468 | |||
GenBank mRNA | AF086315.1 | - | 2, 3 | 126 | ||
AL832080.1 | - | 2, 3 | 136 | |||
AY622964.1 | - | 2, 3 | 121 | |||
AY622965.1 | - | 12, 13 | 1285 | |||
AY789695.1 | - | 12, 13 | 1291 | |||
AY789698.1 | - | 10, 11 | 520 | |||
AY789700.1 | - | 9, 10 | 460 | |||
AY789701.1 | - | 11, 12 | 676 | |||
AY789704.1 | - | 9, 10 | 484 | |||
AY789705.1 | - | 8, 9 | 424 | |||
AY789706.1 | - | 7, 8 | 373 | |||
AY789707.1 | - | 2, 3 | 124 | |||
BC035311.1 | - | 12, 13 | 1462 | 4752921 | ||
BP326188.1 | - | 8, 9 | 485 | |||
CN369399.1 | - | 7, 8 | 395 | |||
EF158320.1 | - | 14, 15 | 1262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
