siRNA Details
Gene Symbol: C1orf158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152290.3 | NP_689503.3 | 4 | 727 | ||
XM_011542434.1 | XP_011540736.1 | 3 | 598 | |||
GenBank mRNA | AK026705.1 | - | 4 | 695 | ||
BC029894.1 | - | 4 | 698 | 5171302 | ||
BX647383.1 | - | 796 |
